Fmoc-D-Abu-OH
Supplier: BACHEM AMERICAS INC
4027306.0001
4027306.0005
B-2920.0001BAEA
309.66
CAD
B-2920.0001BA
B-2920.0005BA
Fmoc-D-Abu-OH
Fmoc-D-Abu-OH
Sequence: Fmoc-D-Abu-OH
Formula:
C₁₉H₁₉NO₄ Storage Temperature: Ambient |
MDL Number:
MFCD00270336 CAS Number: 170642-27-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...