Fmoc-Tyr(PO3(MDPSE)2)-OH

Supplier: BACHEM AMERICAS INC

4026422.0001 4026422.0005
B-2910.0001BAEA 1213.42 CAD
B-2910.0001BA B-2910.0005BA
Fmoc-Tyr(PO3(MDPSE)2)-OH
Fmoc-Tyr(PO3(MDPSE)2)-OH

Sequence: Fmoc-Tyr(PO₃(MDPSE)₂)-OH


Besides a large selection of tyrosine derivatives, Bachem offers derivatives of O-alkylated tyrosines, thyronine, phosphotyrosine and sulfotyrosine. Bachem additionally offers H-[¹⁵N]Tyr-OH, E-3245.
For stable phosphotyrosine mimics see the O-malonyltyrosine derivative B-2825 and the 4-phosphonophenylalanine derivatives B-3895 and A-4725.


Synonym(s): Fmoc-Tyr(PO₃(2-(methyl-diphenyl-silyl)-ethyl)₂)-OH

Formula: C₅₄H₅₄NO₈PSi₂
Storage Temperature: Freezer
MDL Number: MFCD00798641
CAS Number: 158817-11-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR