Fmoc-L-cyclopropylalanine
Supplier: BACHEM AMERICAS INC
Synonyms:
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-cyclopropylpropanoic acid, Fmoc-β-cyclopropyl-L-Alanine, Fmoc-β-cyclopropyl-Ala-OH
4027349.0001
4027349.0005
B-2905.0001BAEA
672.04
CAD
B-2905.0001BA
B-2905.0005BA
Fmoc-L-cyclopropylalanine
Fmoc-L-cyclopropylalanine
Sequence: Fmoc-β-cyclopropyl-Ala-OH
Besides Ala derivatives, Bachem offers a number of derivatives of β-substituted alanines, amongst them: t-Butylalanine Cyanoalanine Cyclohexylalanine (Cha) Cyclopropylalanine 2-Quinolylalanine β-Halogenated Alanines
Formula:
C₂₁H₂₁NO₄ MW: 351.402 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00798640 CAS Number: 214750-76-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...