Fmoc-D-Lys(Alloc)-OH

Supplier: BACHEM AMERICAS INC

Synonyms: Nepsilon-Allyloxycarbonyl-Nα-Fmoc-D-lysine, Fmoc-D-Lys(Aloc)-OH

4027977.0005 4027977.0025
B-2900.0005BAEA 276.63 CAD
B-2900.0005BA B-2900.0025BA
Fmoc-D-Lys(Alloc)-OH
Fmoc-D-Lys(Alloc)-OH

Sequence: Fmoc-D-Lys(Aloc)-OH


For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".

Formula: C₂₅H₂₈N₂O₆
Storage Temperature: Freezer
MDL Number: MFCD00798639
CAS Number: 214750-75-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR