Fmoc-Orn(Alloc)-OH

Supplier: BACHEM AMERICAS INC

Synonyms: Fmoc-Orn(Aloc)-OH, Nδ-Allyloxycarbonyl-Nα-Fmoc-L-ornithine

4026833.0001 4026833.0005
B-2890.0001BAEA 232.35 CAD
B-2890.0001BA B-2890.0005BA
Fmoc-Orn(Alloc)-OH
Fmoc-Orn(Alloc)-OH

Sequence: Fmoc-Orn(Aloc)-OH


Bachem also offers Fmoc-Orn(Boc)-Wang resin (D-1800).

Formula: C₂₄H₂₆N₂O₆
MW: 438.48 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00798637
CAS Number: 147290-11-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR