Fmoc-D-Cys(stbu)-OH
Supplier: BACHEM AMERICAS INC
4026662.0001
4026662.0005
B-2875.0001BAEA
748.67
CAD
B-2875.0001BA
B-2875.0005BA
Fmoc-D-Cys(stbu)-OH
Fmoc-D-Cys(stbu)-OH
Sequence: Fmoc-D-Cys(StBu)-OH
Besides "standard" preloaded resins, Bachem offers laterally anchored Fmoc-cysteine, Fmoc-Cys(SASRIN resin)-OH (D-2170) and resin-linked Fmoc-cysteamine, Fmoc-cysteamine-SASRIN resin (D-2165).
Formula:
C₂₂H₂₅NO₄S₂ Storage Temperature: Freezer |
MDL Number:
MFCD01632050 CAS Number: 501326-55-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...