N,S-Bis-Fmoc-glutathione
Supplier: BACHEM AMERICAS INC
4026654.0250
4026654.1000
B-2855.0250BAEA
401.98
CAD
B-2855.0250BA
B-2855.1000BA
N,S-Bis-Fmoc-glutathione
Proteins and Peptides
Please see also 5-(Pentafluorobenzoylamino)fluorescein (M-2375), a sensitive fluorogenic substrate for the determination of glutathione S-transferase (GST) activity and glutathione (GSH) concentration, and the carba-analog of GSH, ophthalmic acid, H-3145.
Very potent inhibitor of mammalian glyoxalase II (Ki = 0.08 µM for glyoxalase II purified from calf liver).