Fmoc-Dab(Alloc)-OH

Supplier: BACHEM AMERICAS INC

Synonyms: Fmoc-Dab(Aloc)-OH

4026646.0001 4026646.0005
B-2850.0001BAEA 599.86 CAD
B-2850.0001BA B-2850.0005BA
Fmoc-Dab(Alloc)-OH
Fmoc-Dab(Alloc)-OH

Sequence: Fmoc-Dab(Aloc)-OH

Formula: C₂₃H₂₄N₂O₆
MW: 424.453 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00798634
CAS Number: 204316-32-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR