Fmoc-Dap(Alloc)-OH

Supplier: BACHEM AMERICAS INC

4026639.0001 4026639.0005
B-2845.0001BAEA 585.22 CAD
B-2845.0001BA B-2845.0005BA
Fmoc-Dap(Alloc)-OH
Fmoc-Dap(Alloc)-OH

Sequence: Fmoc-Dap(Aloc)-OH


Abbreviated Dap, the abbreviation Dpr can be found in the literature as well. See also Albiziine, Nβ-carbamoyl-Dab, F-3670.
Bachem also offers Fmoc-Dap(Dnp)-SASRIN resin (D-2590) for the SPPS of FRET substrates.

Formula: C₂₂H₂₂N₂O₆
Storage Temperature: Refrigerator
MDL Number: MFCD00273468
CAS Number: 188970-92-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR