Fmoc-n-me-arg(mtr)-oh

Supplier: BACHEM AMERICAS INC

4026642.0001 4026642.0005
B-2840.0001BAEA 820.86 CAD
B-2840.0001BA B-2840.0005BA
Fmoc-n-me-arg(mtr)-oh
Fmoc-n-me-arg(mtr)-oh

Sequence: Fmoc-N-Me-Arg(Mtr)-OH


Bachem additionally offers decarboxy-Arg: Agmatine · sulfate Q-1880.
See also E-2770, H-Arg(Me)-OH · HCl, in the subfamily 'Arginine Substrates and Inhibitors'.

Formula: C₃₂H₃₈N₄O₇S
Storage Temperature: Freezer
MDL Number: MFCD02094400
CAS Number: 214750-72-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR