Fmoc-4-fluoro-L-phenylalanine

Supplier: BACHEM AMERICAS INC

Synonyms: N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-4-fluoro-L-phenylalanine, Fmoc-p-fluoro-Phe-OH, Fmoc-Phe(4-F)-OH, (S)-2-[[[(9H-Fluoren-9-yl)methoxy]carbonyl]amino]-3-(4-fluorophenyl)propanoic acid, N-Fmoc-4-fluoro-L-phenylalanine

4026602.0001 4026602.0005
B-2835.0001BAEA 258.14 CAD
B-2835.0001BA B-2835.0005BA
Fmoc-4-fluoro-L-phenylalanine
Fmoc-4-fluoro-L-phenylalanine

Sequence: Fmoc-p-fluoro-Phe-OH


Bachem offers a considerable choice of ring-substituted phenylalanines, protected derivatives of: p-Aminophenylalanine p-Azidophenylalanine p-Benzoylphenylalanine p-Biphenylalanine p-Bromophenylalanine p-t-Butylphenylalanine p-Carboxyphenylalanine p-Chlorophenylalanine p-Cyanophenylalanine 3,4-Dichlorophenylalanine Fluorophenylalanines p-Iodophenylalanine p-Methylphenylalanine p-Nitrophenylalanine p-Phosphonophenylalanine. p-Methoxy- and p-ethoxyphenylalanine derivatives are listed in the Tyr family, whereas β,β-diphenylalanine derivatives can be found in the "β-Substituted Alanines" family.

Formula: C₂₄H₂₀FNO₄
Storage Temperature: Ambient
MDL Number: MFCD00191197
CAS Number: 169243-86-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR