Fmoc-His(3-Bom)-OH

Supplier: BACHEM AMERICAS INC

4027660.0001 4027660.0005
B-2805.0001BAEA 180.83 CAD
B-2805.0001BA B-2805.0005BA
Fmoc-His(3-Bom)-OH
Fmoc-His(3-Bom)-OH

Sequence: Fmoc-His(3-Bom)-OH


The atoms of the imidazole ring of histidine can be numbered in two different ways. Bachem as well as organic chemists designate the position of N-τ as 1 and the position of N-π as 3, whereas biochemists generally number N-π, the nitrogen atom adjacent to the side chain, as 1, and N-τ as 3.


Synonym(s): Fmoc-His(π-Bom)-OH

Formula: C₂₉H₂₇N₃O₅
Storage Temperature: Freezer
MDL Number: MFCD00077059
CAS Number: 84891-19-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR