Fmoc-Arg(Me)2-OH
Supplier: BACHEM AMERICAS INC
4026426.0001
4026426.0005
B-2745.0001BAEA
1086.93
CAD
B-2745.0001BA
B-2745.0005BA
Fmoc-Arg(Me)2-OH
Fmoc-Arg(Me)2-OH
Sequence: Fmoc-Arg(Me)₂-OH (asymmetrical)
Bachem additionally offers decarboxy-Arg: Agmatine · sulfate Q-1880.
See also E-2770, H-Arg(Me)-OH · HCl, in the subfamily 'Arginine Substrates and Inhibitors'.
Synonym(s): Fmoc-ADMA hydrochloride salt
Formula:
C₂₃H₂₈N₄O₄ Storage Temperature: Freezer |
MDL Number:
MFCD02684510 CAS Number: 268564-10-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...