Fmoc-L-3,3-diphenylalanine
Supplier: BACHEM AMERICAS INC
Synonyms:
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3,3-diphenylpropanoic acid, Fmoc-β,β-diphenyl-Ala-OH
Sequence: Fmoc-β,β-diphenyl-Ala-OH
Besides Ala derivatives, Bachem offers a number of derivatives of β-substituted alanines, amongst them: t-Butylalanine Cyanoalanine Cyclohexylalanine (Cha) Cyclopropylalanine 2-Quinolylalanine β-Halogenated Alanines
Dip derivatives.
Synonym(s): Fmoc-Dip-OH#Fmoc-β-phenyl-Phe-OH#Fmoc-3,3-diphenyl-L-alanine
Formula:
C₃₀H₂₅NO₄ MW: 463.533 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00672337 CAS Number: 201484-50-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...