Fmoc-Glu-OAll

Supplier: BACHEM AMERICAS INC

4026433.0001 4026433.0005
B-2720.0001BAEA 206.51 CAD
B-2720.0001BA B-2720.0005BA
Fmoc-Glu-OAll
Fmoc-Glu-OAll

Sequence: Fmoc-Glu-allyl ester


The derivatives of Glu-NH₂ are assembled in the "Isoglutamine" subfamily.


Synonym(s): Fmoc-Glu-OAll

Formula: C₂₃H₂₃NO₆
Storage Temperature: Freezer
MDL Number: MFCD00467718
CAS Number: 144120-54-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR