Fmoc-Asp-OAll

Supplier: BACHEM AMERICAS INC

4019715.0001 4019715.0005
B-2715.0001BAEA 211.77 CAD
B-2715.0001BA B-2715.0005BA
Fmoc-Asp-OAll
Fmoc-Asp-OAll

Sequence: Fmoc-Asp-allyl ester


Asp-NH₂ is also called isoasparagine, so our Asp-NH₂ derivatives are assembled in the "Isoasparagine" subfamily.


Synonym(s): Fmoc-Asp-OAll

Formula: C₂₂H₂₁NO₆
Storage Temperature: Freezer
MDL Number: MFCD00467715
CAS Number: 144120-53-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR