Fmoc-threoninol

Supplier: BACHEM AMERICAS INC

4026268.0001 4026268.0005
B-2700.0001BAEA 206.51 CAD
B-2700.0001BA B-2700.0005BA
Fmoc-threoninol
Fmoc-threoninol

Sequence: Fmoc-L-threoninol

Formula: C₁₉H₂₁NO₄
Storage Temperature: Freezer
MDL Number: MFCD00270218
CAS Number: 176380-53-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR