(S)-Methyl-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-3-hydroxypropanoate

Supplier: BACHEM AMERICAS INC

Synonyms: (S)-Methyl 2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-3-hydroxypropanoate, Fmoc-Ser-OMe

4026284.0001 4026284.0005
B-2695.0001BAEA 439.41 CAD
B-2695.0001BA B-2695.0005BA
(S)-Methyl-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-3-hydroxypropanoate
(S)-Methyl-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-3-hydroxypropanoate

Sequence: Fmoc-Ser-OMe


Bachem also offers derivatives of phosphoserine and sulfoserine.

Formula: C₁₉H₁₉NO₅
MW: 341.363 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00672334
CAS Number: 82911-78-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR