Fmoc-3-nitro-L-tyrosine

Supplier: BACHEM AMERICAS INC

Synonyms: Fmoc-Tyr(3-NO₂)-OH, Fmoc-3-nitro-Tyr-OH, N-Fmoc-3-nitro-L-tyrosine, Fmoc-Tyr(3-NO2)-OH

4026244.0001 4026244.0005
B-2690.0001BAEA 180.91 CAD
B-2690.0001BA B-2690.0005BA B-2690.0025BA
Fmoc-3-nitro-L-tyrosine
Fmoc-3-nitro-L-tyrosine

Sequence: Fmoc-3-nitro-Tyr-OH


Bachem offers derivatives of (di)halogenated tyrosines, (di)nitrotyrosine and other 3-substituted tyrosines.
See also 3-Methoxytyramine · HCl (Q-1330).


Synonym(s): Fmoc-4-hydroxy-3-nitro-Phe-OH

Formula: C₂₄H₂₀N₂O₇
Storage Temperature: Freezer
MDL Number: MFCD00171383
CAS Number: 136590-09-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR