Fmoc-Lys(biotin)-OH

Supplier: BACHEM AMERICAS INC

4026175.0250 4026175.1000
B-2640.0250BAEA 542.13 CAD
B-2640.0250BA B-2640.1000BA
Fmoc-Lys(biotin)-OH
Fmoc-Lys(biotin)-OH

Sequence: Fmoc-Lys(biotinyl)-OH


For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".
Bachem also offers resins preloaded with Nε-labeled Fmoc-lysine: Fmoc-Lys(biotinyl)-Wang resin (D-2705) and Fmoc-Lys(Dnp)-SASRIN resin (D-2450).


Synonym(s): Fmoc-Lys(Biotin)-OH

Formula: C₃₁H₃₈N₄O₆S
Storage Temperature: Freezer
MDL Number: MFCD00270741
CAS Number: 146987-10-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR