Fmoc-S-2-hydroxyethyl-L-cysteine

Supplier: BACHEM AMERICAS INC

Synonyms: (R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-((2-hydroxyethyl)thio)propanoic acid, Fmoc-Cys(2-hydroxyethyl)-OH

4026001.0001 4026001.0005
B-2630.0001BAEA 748.67 CAD
B-2630.0001BA B-2630.0005BA
Fmoc-S-2-hydroxyethyl-L-cysteine
Fmoc-S-2-hydroxyethyl-L-cysteine

Sequence: Fmoc-Cys(2-hydroxyethyl)-OH


The ω-hydroxy group may be modified, e.g. esterified. S-Hydroxyethylcysteine is formed in blood proteins upon exposure to ethylene oxide. It has been isolated by Kubec et al. from the roots of Petiveria alliacea.

Formula: C₂₀H₂₁NO₅S
Storage Temperature: Freezer
MDL Number: MFCD00672330
CAS Number: 200354-35-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR