Fmoc-Orn-OH·HCl

Supplier: BACHEM AMERICAS INC

4025982.0001 4025982.0005 4025982.0025
B-2625.0001BAEA 185.3 CAD
B-2625.0001BA B-2625.0005BA B-2625.0025BA
Fmoc-Orn-OH·HCl
Fmoc-Orn-OH·HCl

Sequence: Fmoc-Orn-OH · HCl


Bachem also offers Fmoc-Orn(Boc)-Wang resin (D-1800).

Formula: C₂₀H₂₂N₂O₄·HCl
Storage Temperature: Ambient
MDL Number: MFCD00672329
CAS Number: 201046-57-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR