Fmoc-D-Lys(Mtt)-OH
Supplier: BACHEM AMERICAS INC
Synonyms:
Fmoc-Ne-methyltrityl-D-lysine, (R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-6-((diphenyl(p-tolyl)methyl)amino)hexanoic acid, Nα-Fmoc-Nε-(4-methyltrityl)-D-lysine, (2R)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-6-{[(4-methylphenyl)diphenylmethyl]amino}hexanoic acid
4025957.0001
4025957.0005
4025957.0025
B-2620.0001BAEA
370.51
CAD
B-2620.0001BA
B-2620.0005BA
B-2620.0025BA
Fmoc-D-Lys(Mtt)-OH
Fmoc-D-Lys(Mtt)-OH
Sequence: Fmoc-D-Lys(Mtt)-OH
For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".
Formula:
C₄₁H₄₀N₂O₄ Storage Temperature: Freezer |
MDL Number:
MFCD26793593 CAS Number: 198544-94-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...