Fmoc-L-histidine

Supplier: BACHEM AMERICAS INC

Synonyms: Nα-Fmoc-L-histidine, (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(1H-imidazol-4-yl)propanoic acid, Fmoc-His-OH

4013285.0005 4013285.0025
B-2600.0005BAEA 299.43 CAD
B-2600.0005BA B-2600.0025BA
Fmoc-L-histidine
Fmoc-L-histidine

Sequence: Fmoc-His-OH


The atoms of the imidazole ring of histidine can be numbered in two different ways. Bachem as well as organic chemists designate the position of N-τ as 1 and the position of N-π as 3, whereas biochemists generally number N-π, the nitrogen atom adjacent to the side chain, as 1, and N-τ as 3.

Formula: C₂₁H₁₉N₃O₄
MW: 377.4 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00190885
CAS Number: 116611-64-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR