Fmoc-DL-isoser-OH

Supplier: BACHEM AMERICAS INC

4025483.0001 4025483.0005
B-2590.0001BAEA 195.08 CAD
B-2590.0001BA B-2590.0005BA
Fmoc-DL-isoser-OH
Fmoc-DL-isoser-OH

Sequence: Fmoc-DL-Isoser-OH


Bachem also offers derivatives of phosphoserine and sulfoserine.

Formula: C₁₈H₁₇NO₅
Storage Temperature: Ambient
MDL Number: MFCD00672327
CAS Number: 161125-36-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR