Fmoc-Thr(GalNAc(Ac)₃-α-D)-OH

Supplier: BACHEM AMERICAS INC

Synonyms: N-(((9H-Fluoren-9-yl)methoxy)carbonyl)-O-((2S,3R,4R,5R,6R)-3-acetamido-4,5-diacetoxy-6-(acetoxymethyl)tetrahydro-2H-pyran-2-yl)-L-threonine, Fmoc-Thr[GalNAc(Ac)3-α-D]-OH, Fmoc-L-Thr(α-D-GalNAc(Ac)3)-OH

4025798.0025 4025798.0100 4025798.0250
B-2570.0025BAEA 841.57 CAD
B-2570.0025BA B-2570.0100BA B-2570.0250BA
Fmoc-Thr(GalNAc(Ac)₃-α-D)-OH
Fmoc-Thr(GalNAc(Ac)₃-α-D)-OH

Sequence: Fmoc-Thr(GalNAc(Ac)₃-α-D)-OH


Bachem also offers phosphothreonine derivatives.


Synonym(s): N-α-Fmoc-O-β-(2-acetamido-2-deoxy-3,4,6-tri-O-acetyl-α-D-galactopyranosyl)-L-threonine

Formula: C₃₃H₃₈N₂O₁₃
MW: 670.67 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00171374
CAS Number: 116783-35-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR