Fmoc-L-Ser(α-D-GalNAc(Ac)3)-OH
Supplier: BACHEM AMERICAS INC
4025797.0100
4025797.0250
B-2565.0100BAEA
3357.85
CAD
B-2565.0100BA
B-2565.0250BA
Fmoc-L-Ser(α-D-GalNAc(Ac)3)-OH
Fmoc-L-Ser(alpha-D-GalNAc(Ac)3)-OH
Sequence: Fmoc-Ser(GalNAc(Ac)₃-α-D)-OH
Bachem also offers derivatives of phosphoserine and sulfoserine.
Synonym(s): N-α-Fmoc-O-β-(2-acetamido-2-deoxy-3,4,6-tri-O-acetyl-α-D-galactopyranosyl)-L-serine
Formula:
C₃₂H₃₆N₂O₁₃ Storage Temperature: Freezer |
MDL Number:
MFCD00171375 CAS Number: 120173-57-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...