Fmoc-D-Arg(Pbf)-OH

Supplier: BACHEM AMERICAS INC

4025744.0001 4025744.0005 4025744.0025
B-2555.0001BAEA 175.66 CAD
B-2555.0001BA B-2555.0005BA B-2555.0025BA
Fmoc-D-Arg(Pbf)-OH
Fmoc-D-Arg(Pbf)-OH

Sequence: Fmoc-D-Arg(Pbf)-OH


Bachem additionally offers decarboxy-Arg: Agmatine · sulfate Q-1880.

Formula: C₃₄H₄₀N₄O₇S
Storage Temperature: Ambient
MDL Number: MFCD00237010
CAS Number: 187618-60-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR