Fmoc-O-trityl-L-serine

Supplier: BACHEM AMERICAS INC

4020228.0005 4020228.0025 4020228.0100
B-2550.0005BAEA 206.51 CAD
B-2550.0005BA B-2550.0025BA B-2550.0100BA
Fmoc-O-trityl-L-serine
Fmoc-O-trityl-L-serine

Sequence: Fmoc-Ser(Trt)-OH


Bachem also offers derivatives of phosphoserine and sulfoserine.


The trityl group can be removed by 1% TFA, leaving other protecting groups intact.

Formula: C₃₇H₃₁NO₅
Storage Temperature: Freezer
MDL Number: MFCD00153364
CAS Number: 111061-56-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR