Fmoc-O-trityl-L-threonine

Supplier: BACHEM AMERICAS INC

Synonyms: Fmoc-Thr(Trt)-OH, (2S,3R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(trityloxy)butanoic acid

4025725.0005 4025725.0025
B-2545.0005BAEA 424.25 CAD
B-2545.0005BA B-2545.0025BA
Fmoc-O-trityl-L-threonine
Fmoc-O-trityl-L-threonine

Sequence: Fmoc-Thr(Trt)-OH


Bachem also offers phosphothreonine derivatives.


The trityl group can be removed by 1% TFA, leaving other protecting groups intact. On-resin Trt cleavage was employed by Biron et al. in a synthesis of 1,3-oxazole-containing peptides.

Formula: C₃₈H₃₃NO₅
MW: 583.684 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00237034
CAS Number: 133180-01-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR