Fmoc-S-4-methoxytrityl-L-cysteine

Supplier: BACHEM AMERICAS INC

4025726.0001 4025726.0005
B-2540.0001BAEA 593.81 CAD
B-2540.0001BA B-2540.0005BA
Fmoc-S-4-methoxytrityl-L-cysteine
Fmoc-S-4-methoxytrityl-L-cysteine

Sequence: Fmoc-Cys(4-methoxytrityl)-OH
Synonym(s): Fmoc-Cys(Mmt)-OH


Besides "standard" preloaded resins, Bachem offers laterally anchored Fmoc-cysteine, Fmoc-Cys(SASRIN resin)-OH (D-2170) and resin-linked Fmoc-cysteamine, Fmoc-cysteamine-SASRIN resin (D-2165).


4-Methoxytrityl (Mmt) is a very acid-sensitive cysteine protecting group that can selectively be removed by 0.5-1.0% TFA in the presence of scavengers, e.g. TES. Protecting groups of the t-butyl type and S-Trt are left intact under these conditions. Cleavage and cyclization can be achieved concomitantly by adding iodine. It can be used in the solid-phase synthesis of cysteine-containing peptides, and further to selectively deprotect a thiol function in order to introduce a labeling group. Rijkers et al. employed p-methoxytrityl protection for obtaining S-palmitoylated peptides on-resin. The high acid lability of Cys(Mmt) was exploited by Roberts et al. to obtain a library of thioether-linked cyclic peptides. Arendt et al. employed Cys(Mmt) for the synthesis of peptides containing cysteic acid by selective deblocking and oxidation of the thiol moiety with performic acid. Mmt is fully stable to HOBt.

Formula: C₃₈H₃₃NO₅S
Storage Temperature: Freezer
MDL Number: MFCD00270541
CAS Number: 177582-21-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR