Fmoc-Lys(Mtt)-OH

Supplier: BACHEM AMERICAS INC

Synonyms: Nα-[(9H-Fluoren-9-ylmethoxy)carbonyl]-Nepsilon-(4-methyltrityl)-L-lysine, Nα-Fmoc-Nepsilon-(4-methyltrityl)-L-lysine

4025724.0001 4025724.0005 4025724.0025
B-2535.0001BAEA 211.29 CAD
B-2535.0001BA B-2535.0005BA B-2535.0025BA
Fmoc-Lys(Mtt)-OH
Fmoc-Lys(Mtt)-OH

Sequence: Fmoc-Lys(Mtt)-OH


For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".


Mtt can be removed selectively from peptides bound to Wang and Rink resin.

Formula: C₄₁H₄₀N₂O₄
MW: 624.781 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00237166
CAS Number: 167393-62-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR