Fmoc-Lys(palmitoyl)-OH

Supplier: BACHEM AMERICAS INC

4025608.0001 4025608.0005
B-2530.0001BAEA 439.41 CAD
B-2530.0001BA B-2530.0005BA
Fmoc-Lys(palmitoyl)-OH
Fmoc-Lys(palmitoyl)-OH

Sequence: Fmoc-Lys(palmitoyl)-OH


For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".


Synonym(s): Fmoc-Lys(Pam)-OH

Formula: C₃₇H₅₄N₂O₅
Storage Temperature: Freezer
MDL Number: MFCD00237026
CAS Number: 201004-46-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR