Fmoc-O-phospho-L-serine
Supplier: BACHEM AMERICAS INC
Synonyms:
Fmoc-Ser(PO₃H₂)-OH
4025534.0001
4025534.0005
B-2505.0001BAEA
490.51
CAD
B-2505.0001BA
B-2505.0005BA
Fmoc-O-phospho-L-serine
Fmoc-O-phospho-L-serine
Sequence: Fmoc-Ser(PO₃H₂)-OH
Bachem also offers derivatives of phosphoserine and sulfoserine.
N-Terminal phosphoserine residues react with thioesters yielding highly reactive acylphosphates which spontaneously undergo an intramolecular O-N-shift allowing chemical ligation at such positions.
Can be used for solid-phase synthesis of phosphopeptides by the Fmoc strategy.
Formula:
C₁₈H₁₈NO₈P MW: 407.315 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00672324 CAS Number: 158171-15-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...