Fmoc-L-Glu(2-phenylisopropyloxy)-OH

Supplier: BACHEM AMERICAS INC

4025419.0001 4025419.0005
B-2500.0001BAEA 494.16 CAD
B-2500.0001BA B-2500.0005BA
Fmoc-L-Glu(2-phenylisopropyloxy)-OH
Fmoc-L-Glu(2-phenylisopropyloxy)-OH

Sequence: Fmoc-Glu(2-phenylisopropyl ester)-OH


The derivatives of Glu-NH₂ are assembled in the "Isoglutamine" subfamily.


Synonym(s): Fmoc-Glu(OPp)-OH#Fmoc-Glu(O-2-PhiPr)-OH

Formula: C₂₉H₂₉NO₆
Storage Temperature: Freezer
MDL Number: MFCD00672713
CAS Number: 200616-39-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR