Fmoc-Asp(2-phenylisopropyl ester)-OH

Supplier: BACHEM AMERICAS INC

Synonyms: (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-oxo-4-((2-phenylpropan-2-yl)oxy)butanoic acid, Fmoc-Asp(O-2-PhiPr)-OH

4019087.0001 4019087.0005
B-2475.0001BAEA 490.59 CAD
B-2475.0001BA B-2475.0005BA
Fmoc-Asp(2-phenylisopropyl ester)-OH
Fmoc-Asp(2-phenylisopropyl ester)-OH

Sequence: Fmoc-Asp(2-phenylisopropyl ester)-OH


Asp-NH₂ is also called isoasparagine, so our Asp-NH₂ derivatives are assembled in the "Isoasparagine" subfamily.


Synonym(s): Fmoc-Asp(OPp)-OH#Fmoc-Asp(O-2-PhiPr)-OH

Formula: C₂₈H₂₇NO₆
Storage Temperature: Freezer
MDL Number: MFCD00237012
CAS Number: 200336-86-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR