Fmoc-O-phospho-L-tyrosine
Supplier: BACHEM AMERICAS INC
Sequence: Fmoc-Tyr(PO₃H₂)-OH
Besides a large selection of tyrosine derivatives, Bachem offers derivatives of O-alkylated tyrosines, thyronine, phosphotyrosine and sulfotyrosine. Bachem additionally offers H-[¹⁵N]Tyr-OH, E-3245.
For stable phosphotyrosine mimics see the O-malonyltyrosine derivative B-2825 and the 4-phosphonophenylalanine derivatives B-3895 and A-4725.
See Ottinger et al. for coupling conditions. B-2470 forms hydrogels upon enzymatic dephosphorylation. Yang and Xu employed this phenomenon to develop a simple visual assay for detecting inhibitors of enzymes.
Formula:
C₂₄H₂₂NO₈P Storage Temperature: Freezer |
MDL Number:
MFCD00235902 CAS Number: 147762-53-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...