(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)pentanedioic acid

Supplier: BACHEM AMERICAS INC

4025278.0005 4025278.0025
B-2445.0005BAEA 315.24 CAD
B-2445.0005BA B-2445.0025BA
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)pentanedioic acid
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)pentanedioic acid

Sequence: Fmoc-Glu-OH


The derivatives of Glu-NH₂ are assembled in the "Isoglutamine" subfamily.


Simultaneous Nα/α-carboxyl protection can be readily achieved by oxazolidine formation with aldehydes. The product obtained from Fmoc-Glu-OH, paraformaldehyde and TosOH as catalyst has been converted into Dab derivatives, γ-diazomethyl ketones, Nγ-glycosylated Gln, and N'-glycosylated Dab Nγ-ureas.

Formula: C₂₀H₁₉NO₆
Storage Temperature: Ambient
MDL Number: MFCD00237657
CAS Number: 121343-82-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR