Fmoc-Aad(otBu)-OH

Supplier: BACHEM AMERICAS INC

Synonyms: (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-6-(tert-butoxy)-6-oxohexanoic acid

4025264.0001 4025264.0005
B-2440.0005BAEA 3208.4 CAD
B-2440.0005BA B-2440.0001BA
Fmoc-Aad(otBu)-OH
Fmoc-Aad(otBu)-OH

Sequence: Fmoc-Aad(OtBu)-OH
Synonym(s): Fmoc-α-aminoadipic acid-δ-t-butyl ester#(S)-2-(Fmoc-amino)-hexanedioic acid-6-t-butyl ester#Fmoc-Homoglu(OtBu)-OH

Formula: C₂₅H₂₉NO₆
MW: 439.508 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00237006
CAS Number: 159751-47-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR