Fmoc-O-methyl-D-tyrosine

Supplier: BACHEM AMERICAS INC

Synonyms: N-Fmoc-O-methyl-D-tyrosine, Fmoc-D-Tyr(Me)-OH

4025244.0001 4025244.0005
B-2430.0001BAEA 490.51 CAD
B-2430.0001BA B-2430.0005BA
Fmoc-O-methyl-D-tyrosine
Fmoc-O-methyl-D-tyrosine

Sequence: Fmoc-D-Tyr(Me)-OH


Besides a large selection of tyrosine derivatives, Bachem offers derivatives of O-alkylated tyrosines, thyronine, phosphotyrosine and sulfotyrosine. Bachem additionally offers H-[¹⁵N]Tyr-OH, E-3245.
p-Methoxyphenylalanine and p-ethoxyphenylalanine.


Synonym(s): Fmoc-4-methoxy-D-Phe-OH

Formula: C₂₅H₂₃NO₅
MW: 417.461 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00237393
CAS Number: 201335-88-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR