Fmoc-S-trityl-L-homocysteine

Supplier: BACHEM AMERICAS INC

4025118.0001 4025118.0005
B-2405.0001BAEA 856.27 CAD
B-2405.0001BA B-2405.0005BA
Fmoc-S-trityl-L-homocysteine
Fmoc-S-trityl-L-homocysteine

Sequence: Fmoc-Homocys(Trt)-OH


The cysteine homolog is formed by S-demethylation of methionine. Contrary to Cys, Hcy can be incorporated and derivatized without risk of racemization and β-elimination. Base-catalyzed desulfuration of peptides containing a Cys/Hcy mixed disulfide bridge yields cystathionine (carbacystine) peptides.


Synonym(s): (S)-2-(Fmoc-amino)-4-tritylsulfanyl-butyric acid

Formula: C₃₈H₃₃NO₄S
Storage Temperature: Freezer
MDL Number: MFCD00672322
CAS Number: 167015-23-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR