(S)-N-Fmoc-azetidine-2-carboxylic acid

Supplier: BACHEM AMERICAS INC

Synonyms: (S)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)azetidine-2-carboxylic acid, Fmoc-L-azetidine-2-carboxylic acid

4019052.1000 4019052.5000
B-2400.1000BAEA 258.14 CAD
B-2400.1000BA B-2400.5000BA
(S)-N-Fmoc-azetidine-2-carboxylic acid
(S)-N-Fmoc-azetidine-2-carboxylic acid

Sequence: Fmoc-L-azetidine-2-carboxylic acid
Synonym(s): Fmoc-Aze-OH


A suitable derivative for incorporating this strained proline analog in peptides to modify their conformation. Its influence on the stability of the collagen helix has been studied by Zagari et al. Fmoc-Aze-OH was employed in the solid-phase synthesis of potent cyclic GnRH analogs.

Formula: C₁₉H₁₇NO₄
Storage Temperature: Refrigerator
MDL Number: MFCD01320843
CAS Number: 136552-06-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR