Fmoc-N-Me-Glu(OtBu)-OH
Supplier: BACHEM AMERICAS INC
4025071.0001
4025071.0005
B-2395.0001BAEA
828.77
CAD
B-2395.0001BA
B-2395.0005BA
Fmoc-N-Me-Glu(OtBu)-OH
Fmoc-N-Me-Glu(OtBu)-OH
Sequence: Fmoc-N-Me-Glu(OtBu)-OH
The derivatives of Glu-NH₂ are assembled in the "Isoglutamine" subfamily.
Formula:
C₂₅H₂₉NO₆ Storage Temperature: Freezer |
MDL Number:
MFCD00237028 CAS Number: 200616-40-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...