Fmoc-Dap-OH

Supplier: BACHEM AMERICAS INC

4012066.0001 4012066.0005 4012066.0025
B-2385.0001BAEA 264.44 CAD
B-2385.0001BA B-2385.0005BA B-2385.0025BA
Fmoc-Dap-OH
Fmoc-Dap-OH

Sequence: Fmoc-Dap-OH


Abbreviated Dap, the abbreviation Dpr can be found in the literature as well. See also Albiziine, Nβ-carbamoyl-Dab, F-3670.
Bachem also offers Fmoc-Dap(Dnp)-SASRIN resin (D-2590) for the SPPS of FRET substrates.


Educt for the synthesis of Fmoc-β-azidoalanine, which has been incorporated as an IR-probe into peptides.

Formula: C₁₈H₁₈N₂O₄
Storage Temperature: Refrigerator
MDL Number: MFCD00237019
CAS Number: 181954-34-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR