Fmoc-Dap(Boc)-OH

Supplier: BACHEM AMERICAS INC

4018520.0001 4018520.0005 4018520.0025
B-2380.0001BAEA 424.25 CAD
B-2380.0001BA B-2380.0005BA B-2380.0025BA
Fmoc-Dap(Boc)-OH
Fmoc-Dap(Boc)-OH

Sequence: Fmoc-Dap(Boc)-OH


Abbreviated Dap, the abbreviation Dpr can be found in the literature as well. See also Albiziine, Nβ-carbamoyl-Dab, F-3670.
Bachem also offers Fmoc-Dap(Dnp)-SASRIN resin (D-2590) for the SPPS of FRET substrates.

Formula: C₂₃H₂₆N₂O₆
Storage Temperature: Refrigerator
MDL Number: MFCD00235896
CAS Number: 162558-25-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR