Fmoc-Arg(Pbf)-OH
Supplier: BACHEM AMERICAS INC
Synonyms:
Nα-Fmoc-Nω-Pbf-L-arginine, Nα-Fmoc-Nomega-(2,2,4,6,7-pentamethyl-2,3-dihydrobenzo[b]furan-5-ylsulfonyl)-L-arginine, Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-Nω-(2,2,4,6,7-pentamethyldihydrobenzofuran-5-sulfonyl)-L-arginine, Nα-Fmoc-Nomega-(2,2,4,6,7-pentamethyl-2,3-dihydrobenzo[b]furan-5-ylsulphonyl)-L-arginine, Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-Nω-(2,2,4,6,7-pentamethyldihydrobenzofuran-5-sulphonyl)-L-arginine
Sequence: Fmoc-Arg(Pbf)-OH
Bachem additionally offers decarboxy-Arg: Agmatine · sulfate Q-1880.
Pbf protection is especially recommended for multiple-Arg- and Trp-containinng peptides. TFA cleavage is best performed in the presence of a silane scavenger.
Formula:
C₃₄H₄₀N₄O₇S MW: 648.78 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00235804 CAS Number: 154445-77-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...