Fmoc-Arg(Pbf)-OPfp

Supplier: BACHEM AMERICAS INC

4025874.0001 4025874.0005
B-2355.0001BAEA 283.94 CAD
B-2355.0001BA B-2355.0005BA
Fmoc-Arg(Pbf)-OPfp
Fmoc-Arg(Pbf)-OPfp

Sequence: Fmoc-Arg(Pbf)-OPfp


Bachem additionally offers decarboxy-Arg: Agmatine · sulfate Q-1880.

Formula: C₄₀H₃₉F₅N₄O₇S
Storage Temperature: Freezer
MDL Number: MFCD00237040
CAS Number: 200132-16-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR