Fmoc-D-Cha-OH

Supplier: BACHEM AMERICAS INC

4025331.0001 4025331.0025
B-2345.0001BAEA 258.14 CAD
B-2345.0001BA B-2345.0025BA
Fmoc-D-Cha-OH
Fmoc-D-Cha-OH

Sequence: Fmoc-β-cyclohexyl-D-Ala-OH


Besides Ala derivatives, Bachem offers a number of derivatives of β-substituted alanines, amongst them: t-Butylalanine Cyanoalanine Cyclohexylalanine (Cha) Cyclopropylalanine 2-Quinolylalanine β-Halogenated Alanines
Cha corresponds to hydrogenated phenylalanine, the aromatic nucleus is replaced by a more flexible aliphatic cycle.


Synonym(s): Fmoc-D-Cha-OH

Formula: C₂₄H₂₇NO₄
Storage Temperature: Ambient
MDL Number: MFCD00151912
CAS Number: 144701-25-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR