Fmoc-Arg(Boc)₂-OH

Supplier: BACHEM AMERICAS INC

Synonyms: Nomega,Nomega'-Di-Boc-Nalpha-Fmoc-L-arginine, Fmoc-Arg(Boc)2-OH

4018966.0001 4018966.0005
B-2335.0001BAEA 672.04 CAD
B-2335.0001BA B-2335.0005BA
Fmoc-Arg(Boc)₂-OH
Fmoc-Arg(Boc)₂-OH

Sequence: Fmoc-Arg(Boc)₂-OH


Bachem additionally offers decarboxy-Arg: Agmatine · sulfate Q-1880.

Formula: C₃₁H₄₀N₄O₈
MW: 596.68 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00237008
CAS Number: 143824-77-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR