Fmoc-S-trityl-L-penicillamine

Supplier: BACHEM AMERICAS INC

Synonyms: Fmoc-Pen(Trt)-OH, (R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-methyl-3-(tritylthio)butanoic acid

4019778.0001 4019778.0005 4019778.0025
B-2315.0001BAEA 542.13 CAD
B-2315.0001BA B-2315.0005BA B-2315.0025BA
Fmoc-S-trityl-L-penicillamine
Fmoc-S-trityl-L-penicillamine

Sequence: Fmoc-Pen(Trt)-OH


Corresponds to β,β-dimethylcysteine or β-mercaptovaline. For an S-protected derivative of deamino-penicillamine please see Q-1610. The rigidity of disulfide bridges can be increased by replacing Cys by Pen. Pen analogs of enkephalins, RGD-peptides, urotensin and further active peptides with improved potency and/or selectivity have been described. Bachem offers a choice of resins preloaded with Fmoc-Pen derivatives, please see the product family 'Preloaded Resins for SPPS and SPOS'.


Synonym(s): Fmoc-β,β-dimethyl-Cys(Trt)-OH

Formula: C₃₉H₃₅NO₄S
MW: 613.772 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00237388
CAS Number: 201531-88-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR